MAT370170860 Matrix Scientific

[(5-Methyl-1,3,4-oxadiazol-2-yl)methyl]amine

  • MDL number MFCD06739049
  • Cas Number : 125295-22-9
  • Molecular Weight : 113.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/072709.html

Safety Information

MSDS http://www.matrixscientific.com/072709.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart