MAT370170891 Matrix Scientific

4-Boc-Aminonicotinic acid

  • MDL number MFCD03427724
  • Cas Number : 171178-34-0
  • Molecular Weight : 238.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/072740.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/072740.html
Availability details for
Quantity
  • Availability
  • Add to Cart