MAT370171259 Matrix Scientific

9-Amino minocycline hydrochloride

  • MDL number MFCD00919549
  • Cas Number : 149934-21-4
  • Molecular Weight : 508.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 85
Country of Origin USA
Product Link http://www.matrixscientific.com/073110.html

Safety Information

MSDS http://www.matrixscientific.com/073110.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart