MAT370171825 Matrix Scientific

6-Chloropurine-9-riboside

  • MDL number MFCD00005738
  • Cas Number : 2004-06-0
  • Molecular Weight : 302.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/073676.html

Safety Information

MSDS http://www.matrixscientific.com/073676.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart