MAT370172229 Matrix Scientific

6-Trifluoromethyloxindole

  • MDL number MFCD01075220
  • Cas Number : 1735-89-3
  • Molecular Weight : 201.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 184-188
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/074080.html

Safety Information

MSDS http://www.matrixscientific.com/074080.html
pictograms Irritant/Refrigerate
Availability details for
Quantity
  • Availability
  • Add to Cart