MAT370172337 Matrix Scientific

3-Amino-2,2-dimethylpropionamide

  • MDL number MFCD06797568
  • Cas Number : 324763-51-1
  • Molecular Weight : 116.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/074188.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/074188.html
Availability details for
Quantity
  • Availability
  • Add to Cart