MAT370172398 Matrix Scientific

3-Chloro-4-pyridineboronic acid hydrate

  • MDL number MFCD03453058
  • Cas Number : 1256355-22-2
  • Molecular Weight : 175.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.matrixscientific.com/074249.html

Safety Information

MSDS http://www.matrixscientific.com/074249.html
pictograms Irritant/Store in Freezer
Availability details for
Quantity
  • Availability
  • Add to Cart