MAT370172441 Matrix Scientific

3-Amino-4-chlorobenzonitrile

  • MDL number MFCD00017089
  • Cas Number : 53312-79-1
  • Molecular Weight : 152.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 87-91
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/074292.html

Safety Information

hcodes Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart