MAT370172759 Matrix Scientific

4-Chloroimidazole

  • MDL number MFCD09033087
  • Cas Number : 15965-31-8
  • Molecular Weight : 102.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/074610.html

Safety Information

MSDS http://www.matrixscientific.com/074610.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart