MAT370172988 Matrix Scientific

Fmoc-Lys-OH

  • MDL number MFCD00038539
  • Cas Number : 105047-45-8
  • Molecular Weight : 368.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerate
Country of Origin USA
Product Link http://www.matrixscientific.com/074839.html

Safety Information

MSDS http://www.matrixscientific.com/074839.html
pictograms Irritant/Refrigerate
Availability details for
Quantity
  • Availability
  • Add to Cart