MAT370173044 Matrix Scientific

4,6-Dichloropyridin-3-ol

  • MDL number MFCD13190072
  • Cas Number : 1196157-47-7
  • Molecular Weight : 163.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/074895.html

Safety Information

MSDS http://www.matrixscientific.com/074895.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart