MAT370173049 Matrix Scientific

Methyl 1-methyl-1H-[1,2,4]triazole-3-carboxylate

  • MDL number MFCD13191558
  • Cas Number : 57031-66-0
  • Molecular Weight : 141.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/074900.html

Safety Information

MSDS http://www.matrixscientific.com/074900.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart