MAT370173088 Matrix Scientific

2-Iodoxybenzoic acid

  • MDL number MFCD02912492
  • Cas Number : 64297-64-9
  • Molecular Weight : 280.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 250�
Purity 95
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/074939.html

Safety Information

MSDS http://www.matrixscientific.com/074939.html
pictograms Corrosive
Availability details for
Quantity
  • Availability
  • Add to Cart