MAT370173173 Matrix Scientific

Dibutylamine hydrochloride

  • MDL number MFCD00034885
  • Cas Number : 6287-40-7
  • Molecular Weight : 165.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/075024.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/075024.html
Availability details for
Quantity
  • Availability
  • Add to Cart