MAT370173208 Matrix Scientific

2-Bromo-1,3-difluoro-5-methoxybenzene

  • MDL number MFCD00143268
  • Cas Number : 202865-61-0
  • Molecular Weight : 223.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/075059.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/075059.html
Availability details for
Quantity
  • Availability
  • Add to Cart