MAT370173268 Matrix Scientific

2-Adamantanone

  • MDL number MFCD00074737
  • Cas Number : 700-58-3
  • Molecular Weight : 150.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 257�bl.)
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/075119.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/075119.html
Availability details for
Quantity
  • Availability
  • Add to Cart