MAT370173593 Matrix Scientific

2-Nitrophenylhydrazine hydrochloride

  • MDL number MFCD00060205
  • Cas Number : 6293-87-4
  • Molecular Weight : 189.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/075444.html

Safety Information

MSDS http://www.matrixscientific.com/075444.html
pictograms Flammable Solid
Availability details for
Quantity
  • Availability
  • Add to Cart