MAT370173661 Matrix Scientific

Dasatinib

  • MDL number MFCD11046566
  • Cas Number : 302962-49-8
  • Molecular Weight : 488.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/075512.html

Safety Information

msds http://www.matrixscientific.com/075512.html
Availability details for
Quantity
  • Availability
  • Add to Cart