MAT370173763 Matrix Scientific

O-Methylisourea hemisulfate

  • MDL number MFCD00040594
  • Cas Number : 52328-05-9
  • Molecular Weight : 246.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/075614.html

Safety Information

MSDS http://www.matrixscientific.com/075614.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart