MAT370173935 Matrix Scientific

Aminoacetonitrile hydrochloride

  • MDL number MFCD00012850
  • Cas Number : 6011-14-9
  • Molecular Weight : 92.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 166-171�
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/075786.html

Safety Information

hcodes Toxic/Hygroscopic
MSDS http://www.matrixscientific.com/075786.html
Availability details for
Quantity
  • Availability
  • Add to Cart