MAT370173981 Matrix Scientific

tert-Butyl (4R-cis)-6-formaldehydel-2,2-dimethyl-1,3-dioxane-4-acetate

  • MDL number MFCD02093270
  • Cas Number : 124752-23-4
  • Molecular Weight : 258.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 93
Density 1.073
Flash Point 140
Country of Origin USA
Boiling point 320
Product Link http://www.matrixscientific.com/075832.html

Safety Information

MSDS http://www.matrixscientific.com/075832.html
pictograms Irritant/Store in Freezer
Availability details for
Quantity
  • Availability
  • Add to Cart