MAT370174038 Matrix Scientific

2-(Chloromethyl)benzonitrile

  • MDL number MFCD00019745
  • Cas Number : 612-13-5
  • Molecular Weight : 151.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/075889.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/075889.html
Availability details for
Quantity
  • Availability
  • Add to Cart