MAT370174090 Matrix Scientific

2-Methoxy-6-(trifluoromethyl)aniline

  • MDL number MFCD09832384
  • Cas Number : 133863-87-3
  • Molecular Weight : 191.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/075941.html

Safety Information

MSDS http://www.matrixscientific.com/075941.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart