MAT370174157 Matrix Scientific

2-Amino-3-(trifluoromethoxy)benzoic acid

  • MDL number MFCD08669644
  • Cas Number : 561304-41-4
  • Molecular Weight : 221.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/076008.html

Safety Information

MSDS http://www.matrixscientific.com/076008.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart