MAT370174170 Matrix Scientific

2-Bromofuran (stabilized w/copper and layer of water)

  • MDL number MFCD00216609
  • Cas Number : 584-12-3
  • Molecular Weight : 146.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Density 1.662
Flash Point 18.3
Storage temp. Refrigerate/argon/dark
Country of Origin USA
Boiling point 106
Product Link http://www.matrixscientific.com/076021.html

Safety Information

MSDS http://www.matrixscientific.com/076021.html
pictograms Irritant/Light Sensitive/Refrigerate/Stored under Argon
Availability details for
Quantity
  • Availability
  • Add to Cart