MAT370174216 Matrix Scientific

Strontium 2,2'-((5-carboxylato-4-(carboxylatomethyl)-3-cyanothiophen-2-yl)azanediyl)diacetate

  • MDL number MFCD09038742
  • Cas Number : 135459-87-9
  • Molecular Weight : 517.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/076067.html

Safety Information

MSDS http://www.matrixscientific.com/076067.html
pictograms Irritant/Store in Freezer
Availability details for
Quantity
  • Availability
  • Add to Cart