MAT370174417 Matrix Scientific

4-(2-Chloroethyl)morpholine

  • MDL number MFCD00603731
  • Cas Number : 3240-94-6
  • Molecular Weight : 149.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Density 1.11
Storage temp. Freezer/argon
Country of Origin USA
Boiling point 93-94� Torr
Product Link http://www.matrixscientific.com/076268.html

Safety Information

MSDS http://www.matrixscientific.com/076268.html
pictograms Irritant/Stored under Argon/Store in Freezer
Availability details for
Quantity
  • Availability
  • Add to Cart