MAT370174456 Matrix Scientific

3-Ethoxyacrylic acid

  • MDL number MFCD01566841
  • Cas Number : 6192-01-4
  • Molecular Weight : 116.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 109-111
Purity 95
Storage temp. Ambient
Country of Origin USA
Boiling point 87�mHg
Product Link http://www.matrixscientific.com/076307.html

Safety Information

MSDS http://www.matrixscientific.com/076307.html
pictograms Corrosive
Availability details for
Quantity
  • Availability
  • Add to Cart