MAT370174975 Matrix Scientific

4-{2-Methyl-1H-imidazo[4,5-b]pyridin-1-yl}piperidine dihydrochloride

  • MDL number MFCD22373666
  • Cas Number : 1379527-04-4
  • Molecular Weight : 289.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 190-195
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/076646.html

Safety Information

MSDS http://www.matrixscientific.com/076646.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart