MAT371484586 Matrix Scientific

1H-Pyrazole-5-boronic acid

  • MDL number MFCD07368247
  • Cas Number : 376584-63-3
  • Molecular Weight : 111.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/076944.html

Safety Information

MSDS http://www.matrixscientific.com/076944.html
pictograms Irritant/Refrigerate
Availability details for
Quantity
  • Availability
  • Add to Cart