MAT372247619 Matrix Scientific

(4,4'-Difluoro[1,1'-biphenyl]-3-yl)methanamine hydrochloride

  • MDL number MFCD22415175
  • Cas Number : 1401425-38-4
  • Molecular Weight : 255.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/021792.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021792.html
Availability details for
Quantity
  • Availability
  • Add to Cart