MAT372247642 Matrix Scientific

Ethyl 5,5-difluoro-2-oxo-cyclohexanecarboxylate

  • MDL number MFCD20492666
  • Cas Number : 22515-17-9
  • Molecular Weight : 206.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/076966.html

Safety Information

MSDS http://www.matrixscientific.com/076966.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart