MAT372247688 Matrix Scientific

2,2-Dimethylaziridine

  • MDL number MFCD00040452
  • Cas Number : 2658-24-4
  • Molecular Weight : 71.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Boiling point 70-72
Product Link http://www.matrixscientific.com/077022.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/077022.html
Availability details for
Quantity
  • Availability
  • Add to Cart