MAT372247810 Matrix Scientific

5-{[2-Chloro-5-(trifluoromethyl)phenyl]-amino}-5-oxopentanoic acid

  • MDL number MFCD00029870
  • Cas Number : 199803-46-8
  • Molecular Weight : 309.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/077149.html

Safety Information

MSDS http://www.matrixscientific.com/077149.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart