MAT372247891 Matrix Scientific

N-(5-Bromo-4-methyl-1,3-thiazol-2-yl)acetamide

  • MDL number MFCD01924770
  • Cas Number : 21478-95-5
  • Molecular Weight : 235.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/077230.html

Safety Information

MSDS http://www.matrixscientific.com/077230.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart