MAT372247902 Matrix Scientific

5-tert-Butyl-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD00177195
  • Cas Number : 38449-51-3
  • Molecular Weight : 157.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/077241.html

Safety Information

MSDS http://www.matrixscientific.com/077241.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart