MAT372247909 Matrix Scientific

2-(2-Methyl-1H-imidazol-1-yl)ethanol hydrobromide

  • MDL number MFCD20441423
  • Cas Number : 1615-15-2
  • Molecular Weight : 207.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/077248.html

Safety Information

MSDS http://www.matrixscientific.com/077248.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart