MAT372247917 Matrix Scientific

[(5-Methyl-1,3,4-thiadiazol-2-yl)methyl]amine hydrochloride

  • MDL number MFCD20441427
  • Cas Number : 784131-72-2
  • Molecular Weight : 165.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/077256.html

Safety Information

MSDS http://www.matrixscientific.com/077256.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart