MAT372248333 Matrix Scientific

2-[(Methylamino)methyl]benzoic acid

  • MDL number MFCD08756337
  • Cas Number : 527705-23-3
  • Molecular Weight : 165.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/077672.html

Safety Information

MSDS http://www.matrixscientific.com/077672.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart