MAT372248349 Matrix Scientific

(2Z)-2-Chloro-3-phenylacrylic acid

  • MDL number MFCD18432230
  • Cas Number : 705-54-4
  • Molecular Weight : 182.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 137
Country of Origin USA
Product Link http://www.matrixscientific.com/077688.html

Safety Information

MSDS http://www.matrixscientific.com/077688.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart