MAT379353920 Matrix Scientific

2,2'-[3,4-Hexanediylbis(4,1-phenyleneoxy)]-bis(N,N-diethylethanamine) dihydrochloride

  • MDL number MFCD01677853
  • Cas Number : 69-14-7
  • Molecular Weight : 541.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 226-227
Country of Origin USA
Product Link http://www.matrixscientific.com/059778.html

Safety Information

MSDS http://www.matrixscientific.com/059778.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart