MAT379355350 Matrix Scientific

2-Chloro-4-methyl-6-nitroquinoline

  • MDL number MFCD00169016
  • Cas Number : 54965-59-2
  • Molecular Weight : 222.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/079496.html

Safety Information

MSDS http://www.matrixscientific.com/079496.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart