MAT379356062 Matrix Scientific

[2-(1H-Imidazol-1-yl)-1,1-dimethylethyl]amine

  • MDL number MFCD19213768
  • Cas Number : 1265634-26-1
  • Molecular Weight : 139.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/080205.html

Safety Information

MSDS http://www.matrixscientific.com/080205.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart