MAT379356236 Matrix Scientific

{[(1-Ethyl-1H-pyrazol-3-yl)methyl]thio}acetic acid

  • MDL number MFCD09859489
  • Cas Number : 1170645-76-7
  • Molecular Weight : 200.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/080379.html

Safety Information

MSDS http://www.matrixscientific.com/080379.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart