MAT379356401 Matrix Scientific

Ethyl 2-(4-methylbenzyl)-3-oxobutanoate

  • MDL number MFCD13961900
  • Cas Number : 14305-31-8
  • Molecular Weight : 234.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/080544.html

Safety Information

MSDS http://www.matrixscientific.com/080544.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart