MAT379356402 Matrix Scientific

Ethyl 2-(2-methylbenzyl)-3-oxobutanoate

  • MDL number MFCD16317678
  • Cas Number : 104075-51-6
  • Molecular Weight : 234.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Boiling point 120-125�05 Torr
Product Link http://www.matrixscientific.com/080545.html

Safety Information

MSDS http://www.matrixscientific.com/080545.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart