MAT379356403 Matrix Scientific

Ethyl 2-(4-tert-butylbenzyl)-3-oxobutanoate

  • MDL number MFCD16343936
  • Cas Number : 36600-71-2
  • Molecular Weight : 276.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/080546.html

Safety Information

MSDS http://www.matrixscientific.com/080546.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart