MAT379356441 Matrix Scientific

7-Amino-2-methyl[1,2,4]triazolo-[1,5-a]pyrimidin-5(4H)-one

  • MDL number MFCD06613003
  • Cas Number : 5899-94-5
  • Molecular Weight : 165.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/080584.html

Safety Information

MSDS http://www.matrixscientific.com/080584.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart