MAT379356448 Matrix Scientific

Ethyl 2-hydroxy-6-phenylnicotinate

  • MDL number MFCD22374916
  • Cas Number : 58787-26-1
  • Molecular Weight : 243.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/080591.html

Safety Information

MSDS http://www.matrixscientific.com/080591.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart