MAT379356452 Matrix Scientific

6-Amino-1-ethylpyrimidine-2,4(1H,3H)-dione

  • MDL number MFCD00091925
  • Cas Number : 41862-09-3
  • Molecular Weight : 155.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/080595.html

Safety Information

MSDS http://www.matrixscientific.com/080595.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart